Assay Details
Gene Symbol: WRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143857.3 | NP_001137329.3 | 32 - 33 | 4033 | 121 | |
GenBank mRNA | DN531055.1 | - | 3 - 4 | 589 | 121 | |
DN532432.1 | - | 4 - 5 | 433 | 121 | ||
EE354110.1 | - | 7 - 8 | 746 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() |
