Assay Details
Gene Symbol: WRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143857.3 | NP_001137329.3 | 33 - 34 | 4246 | 116 | |
GenBank mRNA | CK976011.1 | - | 1 - 2 | 123 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
