Assay Details
Gene Symbol: FCSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100382.1 | NP_001093852.1 | 3 - 4 | 310 | 85 | |
GenBank mRNA | BC149143.1 | - | 3 - 4 | 310 | 85 | |
BT029847.1 | - | 2 - 3 | 154 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
