Assay Details
Gene Symbol: IFI30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101251.2 | NP_001094721.1 | 2 - 3 | 319 | 56 | |
GenBank mRNA | BC149406.1 | - | 2 - 3 | 320 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
