Assay Details
Gene Symbol: ATRAID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105505.1 | NP_001098975.1 | 3 - 4 | 312 | 86 | |
GenBank mRNA | BC149353.1 | - | 3 - 4 | 312 | 86 | |
BT029884.1 | - | 3 - 4 | 304 | 86 | ||
BT029908.1 | - | 3 - 4 | 304 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
