Assay Details
Gene Symbol: ALG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103346.1 | NP_001096816.1 | 1 - 2 | 45 | 64 | |
GenBank mRNA | BC142132.1 | - | 1 - 2 | 45 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC142132.1, NM_001103346.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
