Assay Details
Gene Symbol: ALG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103346.1 | NP_001096816.1 | 5 - 6 | 885 | 65 | |
GenBank mRNA | BC142132.1 | - | 5 - 6 | 885 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
