Assay Details
Gene Symbol: LARGE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205588.1 | NP_001192517.1 | 10 - 11 | 1457 | 62 | |
GenBank mRNA | BC149956.1 | - | 11 - 12 | 1455 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
