Assay Details
Gene Symbol: LARGE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205588.1 | NP_001192517.1 | 1 - 2 | 103 | 105 | |
GenBank mRNA | BC149956.1 | - | 2 - 3 | 101 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
