Assay Details
Gene Symbol: MARCHF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205812.3 | NP_001192741.1 | 11 - 12 | 986 | 80 | |
GenBank mRNA | BC119893.1 | - | 1 - 2 | 59 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
