Assay Details
Gene Symbol: MARCHF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205812.3 | NP_001192741.1 | 20 - 21 | 1977 | 65 | |
GenBank mRNA | BC119893.1 | - | 10 - 11 | 1050 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
