Assay Details
Gene Symbol: MARCHF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205812.3 | NP_001192741.1 | 2 - 3 | 130 | 106 | |
GenBank mRNA | EV626671.1 | - | 2 - 3 | 130 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
