Assay Details
Gene Symbol: CHRNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192474.1 | NP_001179403.1 | 5 - 6 | 1568 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CHRNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192474.1 | NP_001179403.1 | 5 - 6 | 1568 | 81 |
Set Membership: |
![]() ![]() ![]() ![]() |