Assay Details
Gene Symbol: SNRPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192552.1 | NP_001179481.1 | 1 - 2 | 47 | 73 | |
GenBank mRNA | BC102328.1 | - | 1 - 2 | 46 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102328.1, NM_001192552.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
