Assay Details
Gene Symbol: UNC119B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206287.1 | NP_001193216.1 | 1 - 2 | 243 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: UNC119B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206287.1 | NP_001193216.1 | 1 - 2 | 243 | 111 |
Set Membership: |
![]() ![]() ![]() |