Assay Details
Gene Symbol: CBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206415.1 | NP_001193344.1 | 2 - 3 | 372 | 79 | |
GenBank mRNA | DV828559.1 | - | 2 - 3 | 372 | 79 | |
DV886299.1 | - | 2 - 3 | 308 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
