Assay Details
Gene Symbol: CBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206415.1 | NP_001193344.1 | 4 - 5 | 644 | 94 | |
GenBank mRNA | DV886299.1 | - | 4 - 5 | 580 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
