Assay Details
Gene Symbol: TMED4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206433.1 | NP_001193362.1 | 1 - 2 | 202 | 96 | |
GenBank mRNA | BC123580.1 | - | 1 - 2 | 203 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
