Assay Details
Gene Symbol: TMED4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206433.1 | NP_001193362.1 | 2 - 3 | 304 | 65 | |
GenBank mRNA | BC123580.1 | - | 2 - 3 | 305 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
