Assay Details
Gene Symbol: PCBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206474.1 | NP_001193403.1 | 1 - 2 | 120 | 90 | |
GenBank mRNA | BC103434.1 | - | 1 - 2 | 122 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001206474.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
