Assay Details
Gene Symbol: AASDHPPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206477.1 | NP_001193406.1 | 3 - 4 | 604 | 100 | |
GenBank mRNA | BC102625.1 | - | 3 - 4 | 573 | 100 | |
DT813625.1 | - | 3 - 4 | 526 | 100 | ||
DT860683.1 | - | 3 - 4 | 605 | 100 | ||
EV716630.1 | - | 3 - 4 | 563 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
