Assay Details
Gene Symbol: CHRNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192710.2 | NP_001179639.1 | 6 - 7 | 1615 | 54 | |
GenBank mRNA | BC119878.1 | - | 6 - 7 | 1774 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
