Assay Details
Gene Symbol: RAB11FIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192771.1 | NP_001179700.1 | 3 - 4 | 1551 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RAB11FIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192771.1 | NP_001179700.1 | 3 - 4 | 1551 | 75 |
Set Membership: |
![]() ![]() ![]() |