Assay Details
Gene Symbol: PAK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206594.2 | NP_001193523.2 | 4 - 5 | 1313 | 73 | |
GenBank mRNA | EE224118.1 | - | 1 - 2 | 568 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
