Assay Details
Gene Symbol: LIPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206597.1 | NP_001193526.1 | 4 - 5 | 577 | 91 | |
GenBank mRNA | DV916511.1 | - | 2 - 3 | 267 | 91 | |
DV917166.1 | - | 5 - 6 | 559 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
