Assay Details
Gene Symbol: STIL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206662.2 | NP_001193591.1 | 11 - 12 | 2213 | 71 | |
GenBank mRNA | CK776513.1 | - | 4 - 5 | 722 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
