Assay Details
Gene Symbol: TTPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206676.1 | NP_001193605.1 | 4 - 5 | 662 | 101 | |
GenBank mRNA | GH315554.1 | - | 3 - 4 | 375 | 101 | |
LC223005.1 | - | 4 - 5 | 697 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
