Assay Details
Gene Symbol: BCAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206698.2 | NP_001193627.1 | 4 - 5 | 963 | 108 | |
GenBank mRNA | BC114070.1 | - | 4 - 5 | 964 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
