Assay Details
Gene Symbol: TNFRSF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184950.1 | NP_001171879.1 | 1 - 2 | 151 | 109 | |
GenBank mRNA | FJ959390.1 | - | 1 - 2 | 151 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
