Assay Details
Gene Symbol: ISY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191173.2 | NP_001178102.1 | 9 - 10 | 794 | 90 | |
GenBank mRNA | BC142461.1 | - | 9 - 10 | 769 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
