Assay Details
Gene Symbol: GCSAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191208.1 | NP_001178137.1 | 5 - 6 | 262 | 99 | |
GenBank mRNA | DY134218.1 | - | 5 - 6 | 269 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
