Assay Details
Gene Symbol: UST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206826.1 | NP_001193755.1 | 3 - 4 | 457 | 76 | |
NM_001206827.1 | NP_001193756.1 | 3 - 4 | 457 | 76 | ||
GenBank mRNA | DV888180.1 | - | 1 - 2 | 108 | 76 | |
DV895087.1 | - | 2 - 3 | 198 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
