Assay Details
Gene Symbol: HRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242346.1 | NP_001229275.1 | 4 - 5 | 613 | 77 | |
GenBank mRNA | DN532777.1 | - | 2 - 3 | 271 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
