Assay Details
Gene Symbol: LOC508666 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244199.1 | NP_001231128.1 | 2 - 3 | 469 | 106 | |
GenBank mRNA | BC110158.1 | - | 2 - 3 | 439 | 106 | |
EV633346.1 | - | 2 - 3 | 472 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
