Assay Details
Gene Symbol: IGFBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174554.3 | NP_776979.2 | 1 - 2 | 453 | 100 | |
GenBank mRNA | BC111197.1 | - | 1 - 2 | 454 | 100 | |
X54979.1 | - | 445 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
