Assay Details
Gene Symbol: LPAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192235.1 | NP_001179164.1 | 2 - 3 | 228 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001192235.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
