Assay Details
Gene Symbol: TANK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192264.1 | NP_001179193.1 | 2 - 3 | 186 | 81 | |
GenBank mRNA | BC119873.1 | - | 2 - 3 | 196 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
