Assay Details
Gene Symbol: ZNF384 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192394.1 | NP_001179323.1 | 2 - 3 | 293 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001192394.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
