Assay Details
Gene Symbol: MRPL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192395.1 | NP_001179324.1 | 3 - 4 | 412 | 84 | |
GenBank mRNA | BC104520.1 | - | 3 - 4 | 364 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
