Assay Details
Gene Symbol: SDCCAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193109.2 | NP_001180038.2 | 2 - 3 | 371 | 81 | |
GenBank mRNA | EV626017.1 | - | 2 - 3 | 333 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
