Assay Details
Gene Symbol: DLEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193198.3 | NP_001180127.3 | 8 - 9 | 1418 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DLEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193198.3 | NP_001180127.3 | 8 - 9 | 1418 | 96 |
Set Membership: |
![]() ![]() ![]() ![]() |