Assay Details
Gene Symbol: TNNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034351.2 | NP_001029523.1 | 4 - 5 | 350 | 62 | |
GenBank mRNA | BC102995.1 | - | 4 - 5 | 351 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
