Assay Details
Gene Symbol: ASMTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035058.1 | NP_001030230.1 | 12 - 13 | 1654 | 81 | |
GenBank mRNA | BC102999.1 | - | 12 - 13 | 1654 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
