Assay Details
Gene Symbol: RGS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046600.2 | NP_001040065.1 | 3 - 4 | 334 | 114 | |
GenBank mRNA | BC114106.1 | - | 3 - 4 | 336 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
