Assay Details
Gene Symbol: MIR2310 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031046.1 | 1 | 30 | 60 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
MYC proto-oncogene, bHLH transcription factor |
MYC | CMYC | -731.3kb | Anti-sense | Chr. 14 - 12653171 - 12659616 on NCBI Build ARS-UCD1.2 |
Downstream | LRAT domain containing 2 | LRATD2 | FAM84B | 529.4kb | Sense | Chr. 14 - 13920349 - 13925812 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
