Assay Details
Gene Symbol: RNASEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_130175.1 | 2 - 3 | 254 | 92 | ||
GenBank mRNA | EH143926.1 | - | 2 - 3 | 208 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
arachidonate 12-lipoxygenase, 12S type |
ALOX12 | 12-LOX | -2.5kb | Sense | Chr. 19 - 26786538 - 26800876 on NCBI Build ARS-UCD1.2 |
Downstream | chromosome 19 C17orf49 homolog | C19H17orf49 |
BAP18, C17orf49 |
901.0 | Sense | Chr. 19 - 26804519 - 26807430 on NCBI Build ARS-UCD1.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
