Assay Details
Gene Symbol: GPRC5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272010.1 | NP_001258939.1 | 1 - 2 | 58 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001272010.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
