Assay Details
Gene Symbol: CBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY962100.1 | - | 3 - 4 | 172 | 67 | |
BT029790.1 | - | 1 - 2 | 516 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY962100.1 | - | 3 - 4 | 172 | 67 | |
BT029790.1 | - | 1 - 2 | 516 | 67 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |