Assay Details
Gene Symbol: hmr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026479.4 | NP_001021650.1 | 15 - 16 | 4058 | 89 | |
GenBank mRNA | AJ307058.1 | - | 15 - 16 | 4058 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
