Assay Details
Gene Symbol: gly-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059473.8 | NP_491874.1 | 2 - 3 | 299 | 129 | |
GenBank mRNA | AF154122.1 | - | 2 - 3 | 299 | 129 | |
AY037802.1 | - | 6 - 7 | 682 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
